missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek 500 Labo Coverall
GREENER_CHOICE

Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ CHF7 Hooded Laboratory Coveralls are robust yet lightweight with a 3-piece hood and gusset for improved fit.

Brand:  DuPont™ D14529877

231.70 EUR valid until 2025-12-31
Use promo code "24380" to get your promotional price.


 View more versions of this product

Product Code. 11733113

  • 289.00 € / Pack of 25
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Excellent barrier against fine particles and fibres
  • Ultra-low-linting and antistatically treated
  • Tyvek zipper and zipper flap for enhanced protection
  • Composed of flash spun high density polyethylene
  • Elasticated face, wrists and ankles as well as glued-in elastic waist
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols
  • Applications include the pharmaceutical industry, laboratories, cosmetics, optical and electronics
  • Stitched internal seams to help reduce contamination from the inside to the outside of the garment
  • Attached overshoes consisting of garment material with friction-coated sole to provide added slip resistance
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hooded Coveralls
Tyvek
Elastic
Elastic
3-panel
Stitched, Taped
Zipper flap, tunneled cuff and facial elastics
2X-Large
White
Elastic
Hooded
Zipper with Storm Flap
Category III, Type 5 and 6, EN 1073-2
116 to 124 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tyvek 500 Labo Coverall > Size: 2X-Large

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.