Learn More
ABEBA™ Uni6 White Occupational Sneakers O1
ABEBA™ Uni6 White Occupational Sneakers O1 made of water repellent knitted textile with a slip resistant PU/TPU outsole.
Brand:  ABEBA™ 36772/35
 View more versions of this product
Product Code. 16358141
  • 118.00 € / 1 pair
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Breathability, lightweight, comfort
  • Comply to EN ISO 20347 : 2012, O1 FO, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water repellent and breathable knitted textile
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable Active Comfort insole
Specifications
Specifications
CE EN ISO 20347 :2012, O1, FO, SRC, ESD
White
Breathable silver ions lining
35
Polyurethane, Thermoplastic Polyurethane
1 Pair
Laces
Stainless steel
Textile
Slip and abrasion resistant, ESD dissipative, shock absorbent
Sneakers
EN ISO 20347 : 2012; DIN EN 61340
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
ABEBA™ Uni6 White Occupational Sneakers O1 > Size: 35