Learn More
Cole-Parmer™ UV Tubes For Ultraviolet Equipment
Use these replacement UV tubes with ultraviolet lamps.
Brand:  Cole-Parmer™ 09815-36
 View more versions of this product
Product Code. 11800390
Description
Description
  • Replacement Ultraviolet Tubes
  • Product may contain trace amounts of mercury
  • Use caution with breakage, cleanup, shipping and/or disposal
Specifications
Specifications
UV-C
254 nm
4 W, 254 nm lamps
UV Tube
4 W
15.5 mm
134.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ UV Tubes For Ultraviolet Equipment > Diameter: 15.5mm; Wavelength: 254nm; Wattage: 4w