missing translation for 'onlineSavingsMsg'
Learn More

UVP Replacement UV Tubes

For use with 8w UV lamps.

26.45 € - 74.00 €

Specifications

Wattage 8 W
Product Type Replacement UV Tube
View More Specs

Products 3
Product Code Brand For Use With (Equipment) Wavelength Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Wavelength Price Quantity & Availability  
12862989
View Documents
UVP
34-0006-01
Model TLW-20 transilluminator, model CL-1000 crosslinker, models 2UV and 3UV transilluminator and EL series UV lamp 365 nm
26.45 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
 
12822989
View Documents
UVP
34-0007-01
CL-1000 crosslinker, DE-50 eprom eraser, models 2UV and 3UV transilluminator and EL series UV lamp 254 nm
49.80 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11798231
View Documents
UVP
34-0042-01
CL-1000M crosslinker, TMW-20 and benchtop series transilluminator and EL series UV lamp 302 nm
74.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

For use with 8w UV lamps.

  • For use with 8w UV lamps
Specifications

Specifications

8 W
Replacement UV Tube
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
UVP Replacement UV Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.