Learn More
UVP Replacement UV Tubes
For use with 8w UV lamps.
Brand:  UVP 34-0007-01
 View more versions of this product
Product Code. 12822989
  • 49.80 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
For use with 8w UV lamps.
  • For use with 8w UV lamps
Specifications
Specifications
8 W
CL-1000 crosslinker, DE-50 eprom eraser, models 2UV and 3UV transilluminator and EL series UV lamp
28.7 cm
254 nm
11.33 in.
Replacement UV Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
UVP Replacement UV Tubes > Shortwave 254nm