Learn More
UVP Replacement UV Tubes
For use with 8w UV lamps.
Brand:  UVP 34-0042-01
 View more versions of this product
Product Code. 11798231
  • 74.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
For use with 8w UV lamps.
  • For use with 8w UV lamps
Specifications
Specifications
8 W
302 nm
CL-1000M crosslinker, TMW-20 and benchtop series transilluminator and EL series UV lamp
Replacement UV Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
UVP Replacement UV Tubes > Midrange 302nm