Learn More
Fisherbrand™ Vacuum Regulator with Dial Gauge
Designed for use with Fisherbrand Chemical Duty Diaphragm Vacuum Pumps. Fisherbrand™ Vacuum Regulators feature regulator valve for intake that allows the adjustment of the ultimate pressure.
Brand:  Fisherbrand™ 700458-10
Product Code. 15549632
  • 253.76 € / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With DN8 hose nozzle for 8mm I.D. vacuum hose
  • Regulator valve for intake allows the adjustment of the ultimate pressure
  • Dial gauge indicates the vacuum level
  • Air regulator valve mounted directly on the pump
  • For Chemical Duty Diaphragm Vacuum Pump Cat. No 10287097
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For Fisherbrand Chemical Duty Diaphragm Vacuum Pump, Cat. No 10287097
with DN8 hose nozzle for 8mm ID vacuum hose
Model DBR-B Vacuum Regulator with Dial Gauge
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Vacuum Regulator with Dial Gauge >