Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system
Brand:  Cole-Parmer™ 12018-85
 View more versions of this product
Product Code. 15844594
  • 481.00 € / Each
Estimated Shipment: 20-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Select from a variety of cap sizes to fit your container
  • Different materials, numbers and sizes of ports help customize to your application
Services
None
Specifications
Specifications
Solvent Safety
PTFE
1/2 in. NPT(F), 1/4 in. NPT(F)
Solvent-Compatible Containers
DIN 45
Solvent Waste Manifold, PTFE, DIN 45, 1 x 1/2 in. NPTF, 3 x 1/4 in. NPTF; 1/ea
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys > PTFE, DIN 45, 1x1/2 in NPTF, 3x1/4 in NPTF; 1/ea