Learn More
Product Code | Brand | Model | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Includes | Price | Quantity & Availability | |||||
17825666
|
3M™
7100258667 |
Comfort Faceseal |
320.00 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
17855666
|
3M™
7100256071 |
Flame Resistant Poly Faceseal |
313.00 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
17835666
|
3M™
7100257937 |
Comfort Faceseal |
515.00 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
17809271
|
3M™
7100257936 |
Flame Resistant Faceseal |
515.00 €
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
11783916
|
3M™
7000104037 |
M-406 | Highly Durable Shroud |
825.00 €
Each |
Estimated Shipment: 26-05-2025
Log in to see stock. |
|||||
17815666
|
3M™
7100258668 |
Flame Resistant Shroud |
725.00 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
Description
- M-Series are specifically designed to work together with your 3M™ Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
- Deflector allows users to direct the airflow inside the headtop to increase comfort.
- Fully-adjustable suspension for customized fit and weight distribution.
- Accessory options availalble for protection to help handle the environments you encounter.
- Visor combines good peripheral and downward vision.
- Specific headtops can be used with a range of 3M™ Peltor™ hearing protection as well as an attachable microphone for ease of communication.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
3M™ Versaflo™ M-Series Faceshields and Helmets