Learn More
Ansell™ VersaTouch™ 37-501 Series Blue Nitrile Immersion Gloves
Resists many solvents and oil based chemicals. Ansell™ VersaTouch™ 37-501 Series Blue Nitrile Immersion Gloves have good abrasion resistance.
64.45 €
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15324092
|
Ansell™
37501105 |
10.5 |
64.45 €
12 pairs |
In Stock
Log in to see stock. |
|||||
15334082
|
Ansell™
37501065 |
6.5 |
64.45 €
12 pairs |
In Stock
Log in to see stock. |
|||||
15354082
|
Ansell™
37501075 |
7.5 |
64.45 €
12 pairs |
In Stock
Log in to see stock. |
|||||
15394082
|
Ansell™
37501095 |
9.5 |
64.45 €
12 pairs |
In Stock
Log in to see stock. |
|||||
15374082
|
Ansell™
37501085 |
8.5 |
64.45 €
12 pairs |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
Description
- Excellent resistance to many solvents and oil based chemicals
- Very good abrasion resistance
- Flexible and comfortable for an excellent dexterity
- Gauntlet cuff style
- Raised diamond grip
- Powder free
Specifications
0.42 mm | |
Gauntlet | |
37-501 |