missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Vial Rack

This vial rack can be used for all vials (11.6 x 32 mm) with flat bottom. It is a 50 position vial rack, made from blue, chemically resistant polypropylene. Safe standing position of the vials on the lab bench. They feature an alpha-numerical index and are stackable when empty.

Brand:  Macherey-Nagel™ 702502

14.18 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15798289

  • 17.12 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Holds 50 vials
  • Blue polypropylene
  • Chemically resistant
  • Alph-numeric grid
  • Stackable when empty
  • Ideal for storage or transportation
Specifications

Specifications

Rack
Blue
Yes, When Empty
190 x 100 x 22 mm
Vial Rack
50
50 of 12mm (Max.) Diameter Vials
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Vial Rack > Vial Rack; Positions: 50; Color: Blue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.