Learn More
Thermo Scientific™ Violet Red Bile Glucose Agar (EP, USP, JP, BP)
Selectively enumerate Enterobacteriaceae in food, animal feed, and environmental samples with this ready-to-use medium.
Brand: Thermo Scientific™ PO1167A
11.23 EUR valid until 2025-12-31
Use promo code "24364" to get your promotional price.
Description
Selectively enumerate Enterobacteriaceae in food, animal feed and environmental samples with ready-to-use Thermo Scientific™ Oxoid™ Violet Red Bile Glucose (VRBG) Agar (EP, USP, JP, BP) while adhering to ISO 21528 and ISO 11133:2014. Due to the careful addition of selective inhibitory components, neutral red, crystal violet, and bile salts, the medium provides selectivity against non-target Gram-positive bacteria. The medium is tested in accordance with International Pharmacopoeia1,2,3,4.
Use Violet Red Bile Glucose (VRBG) Agar (EP, USP, JP, BP) for examination of food, animal feed, and environmental samples, for the presence of indicator groups such as coliforms. As well as providing evidence of food hygiene and safety, indicator groups provide a measure of food quality and spoilage potential. One of the most common groups of bacteria employed as indicator organisms by the food industry are the coliforms, which can be regarded as a subgroup within the Enterobacteriaceae. However, in an attempt to improve assessment of food safety and quality by the food industry, there has been a move towards testing for total Enterobacteriaceae rather than limiting testing to coliforms.
In processed foods, particularly foods subjected to heat treatment, these organisms can provide a reliable indication of process failure, under-processing, or post-processing contamination.
- Easy-to-read results: due to inclusion of neutral red indicator and crystal violet in the medium Enterobacteriaceae form purple pink colored colonies. Adheres to ISO 21528 and ISO 11133:2014 standards.
- Conforms to formulations detailed in International Pharmacopoeia
- Ready-to-use convenience of prepared media.
Mossel5 modified Violet Red Bile Lactose Agar by adding glucose to improve recovery of all Enterobacteriaceae. Later work by Mossel and Cowell6,7 demonstrated that lactose could be omitted, resulting in the formulation known as VRBGA.
Enterobacteriaceae rapidly ferment glucose and so reduce the pH of the medium, which produces purple/pink coloured colonies due to the inclusion of the neutral red indicator and crystal violet in the medium. These colonies are usually surrounded by purple halos of precipitated bile salts. Crystal violet and bile salts inhibit the growth of Gram-positive flora.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Violet / Red, clear | |
Violet Red Bile Glucose Agar (EP, USP, JP, BP) | |
United Kingdom | |
18.5g – 20.5g | |
10/Pk. |
2°C to 10°C | |
90mm Plate | |
Agar | |
Flow Wrapped - Single Layer |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4500106 | Certificate of Analysis | 28/03/2025 | PO1167A |
4500041 | Certificate of Analysis | 24/03/2025 | PO1167A |
4497510 | Certificate of Analysis | 15/03/2025 | PO1167A |
4496392 | Certificate of Analysis | 09/03/2025 | PO1167A |
4495911 | Certificate of Analysis | 07/03/2025 | PO1167A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.