Learn More
Gibco™ VP-SFM (1X)

VP-SFM is a serum-free, ultra-low protein (5 μg/mL) medium containing no proteins, peptides, or other components of animal or human origin. It is designed for the growth of VERO cells for virus production.
Brand: Gibco™ 11681020
113.40 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
VP-SFM is a serum-free, ultra-low protein (5μg/mL) medium containing no proteins, peptides, or other components of animal or human origin. It is designed for the growth of VERO cells for virus production (1). This medium is suitable for the growth of COS-7, MDCK, BHK-21 (suspension culture) and HEp2 cells, and for recombinant protein production.
Formulated without L-glutamine for increased stability; add 20 mL of 200 mM L-glutamine or 20 mL of GlutaMAX™-I supplement per 1,000 mL of medium prior to use.

Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Animal Origin-free, Serum-free | |
Suspension Cell Culture | |
VP Serum Free Medium (SFM) | |
No Glutamine | |
Bottle(s) | |
Gibco™ | |
Room Temperature |
MDCK, VERO, HEp2, BHK-21, COS-7 | |
1 X | |
Liquid | |
Phenol Red, Sodium Pyruvate, Sodium Bicarbonate | |
Low | |
No antibiotics | |
1 L |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2730659 | Certificate of Analysis | 29/01/2025 | 11681020 |
2458461 | Certificate of Analysis | 29/01/2025 | 11681020 |
2492803 | Certificate of Analysis | 29/01/2025 | 11681020 |
2360185 | Certificate of Analysis | 29/01/2025 | 11681020 |
2472366 | Certificate of Analysis | 29/01/2025 | 11681020 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.