Learn More
Thermo Scientific™ Water Softener Accessories
Remove hard ions, such as calcium and magnesium, using these accessories. Softened water extends the life of the reverse osmosis membrane.
Brand: Thermo Scientific™ 06.1000
Description
Remove hard ions, such as calcium and magnesium, using Thermo Scientific™ Water Softener Accessories. Softened water extends the life of the reverse osmosis membrane.
Sensor controller for water softening monitoring
- The Sensor controller SE is a monitoring system for a quality-controlled operation of the water softening system
Total hardness measurement (titration solution)
- Necessary to determine the feed water hardness and calculate the softener program settings
Bacteria guard for water softeners
- For disinfecting the ion exchange bed during each regeneration
- The bacteria guard extracts chlorine from a part of the salt water to disinfect the ion exchange resin
Flexible connecting hose 1in. with stainless steel jacketing, length 1 meter
- Smart and easy connection of your Thermo Scientific Water softener to wall connectors
- Alternative instead of fixed piping, which offers more flexibility for maintenance
Mounting block 1in. with blending valve
- For comfortable bypassing of the softener for maintenance, reparations, etc
Blending valve OVP for 1in. softening systems for permanent hardness adjustment
- If a specific water hardness is required after the softener, the blending valve will some hard water into the soft water, coming out of the softeners output
Blending valve OVP for 1 1/2in. softening systems for permanent hardness adjustment
- If a specific water hardness is required after the softener, the blending valve will some hard water into the soft water, coming out of the softeners output
Specifications
Hardness Test Kit | |
Cabinet Softening System WAE, Double Softening System WAT, Double Softening System WMS | |
Hardness Test Kit |
Product Suggestions
Customers who viewed this item also viewed.