Learn More
Cytiva Whatman™ pH Indicator Papers, CF Strips
Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
Brand:  Cytiva 2613-991
22.08 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 10377481
  • 29.30 € / Pack of 100
Edge
Explore more special offers
Description
Description
Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
  • Plastic supports that retain stiffness in liquids
  • Paper segments on each strip have chemically bonded dyes; can't leach into solution
  • Complementary indicator charts provide clear comparison
  • Full-range strips (0 to 14) graduated in 1pH units; wide-range strips (4.5 to 10pH) in 0.5pH units
  • Packs of 100; each strip is approximately 0.25 × 3 in. (6 × 80mm)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0 to 14
1 pH
100 Pack
80 x 6 mm
6 mm
Strip
Plastic supports, paper segments on each strip, complementary indicator charts
pH Test Strip, Color Bonded
80 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Whatman™ pH Indicator Papers, CF Strips > CF strips; Range: pH 0 to 14