missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ UNIPLATE™ Microplates

Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).

315.00 € - 439.00 €

Specifications

Format Standard
Description UNIPLATE microplates
Color Natural
Material Polypropylene
For Use With (Application) Ideal for homogeneous assay techniques utilized in high-throughput screening
View More Specs

Products 3
Product Code Brand Sterility Volume (Metric) Well Well Shape No. of Wells Price Quantity & Availability  
Product Code Brand Sterility Volume (Metric) Well Well Shape No. of Wells Price Quantity & Availability  
11378674
View Documents
Cytiva
7701-5102
Non-sterile 10 mL Round 24
439.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
 
11328684
View Documents
Cytiva
7701-5500
Non-sterile 5 mL Flat Bottom (F-well) 48
315.00 €
Pack of 25
In Stock
to see stock.
Add to basket
 
10423772
View Documents
Cytiva
7701-5110
Sterile 10 mL Round 24
382.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).

  • Withstand low temperatures (-80°C) for long-term storage
  • Conform to ANSI™/SBS standards for compatibility with most microplate readers and automated plate-handling devices
Specifications

Specifications

Standard
Natural
Ideal for homogeneous assay techniques utilized in high-throughput screening
UNIPLATE microplates
Polypropylene
25/Pk.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Whatman™ UNIPLATE™ Microplates

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.