missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ UNIPLATE™ Microplates

Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).

Brand:  Cytiva 7701-5102

346.69 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.


 View more versions of this product

Product Code. 11378674

  • 439.00 € / Pack of 25
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).

  • Withstand low temperatures (-80°C) for long-term storage
  • Conform to ANSI™/SBS standards for compatibility with most microplate readers and automated plate-handling devices
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
4 x 6
Polypropylene
24
Round
25/Pk.
UNIPLATE microplates
Natural
10 mL
Non-sterile
Ideal for homogeneous assay techniques utilized in high-throughput screening
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Whatman™ UNIPLATE™ Microplates > Round well; Well volume: 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.