Learn More
DWK Life Sciences Wheaton™ 85mm Condenser
For extraction of analytes
Brand:  DWK Life Sciences 415501
Product Code. 12401571
  • 159.00 € / Each
Estimated Shipment: 28-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 24-400 bottom screw thread
  • 13-425 side screw thread
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USP Type I and ASTM E 438 Type I, Class A requirements
3.34 in.
Borosilicate Glass
Used for continuous extraction of analytes from a solid into an organic solvent
85 mm
Condenser
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ 85mm Condenser > 24-400 bottom screw thread; 13-425 side screw thread