missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Serum Vials

Available as clear and amber vials

Brand:  DWK Life Sciences 223683

 View more versions of this product

Product Code. 10472752

This item is not returnable. View return policy

Description

Description

Available as clear and amber vials

Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.

  • Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
  • Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
  • Withstand extreme temperature variations
  • Necks finished for aluminum crimp seals
  • Blowback feature of neck and specially designed bottom radius for lyophilization applications
  • Packaged in shrink-wrapped modules to reduce particulate contamination

Lyophilization, packaging, and sampling

Compliance

Low extractable borosilicate glass meets ASTM™ Type 1, Class A and USP Type 1 standards

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.27 in.
Clear
Borosilicate Glass
For lyophilization, packaging, sampling and freeze drying applications
Wheaton
Autoclavable
32 mm
Serum Vial
7 mm
15 mm
2 mL
0.5 dr.
2 mL
Crimp Cap
ASTM E438, USP Type I, Type I, Class A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Serum Vials > Clear; 2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.