missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ White Disposable Polypropylene Coveralls, 4500 Series
Disposable polypropylene coveralls offer Category I protection for non-hazardous applications.
120.00 €
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Product Code | Brand | Size | Height (English) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Height (English) | Height (Metric) | Price | Quantity & Availability | |||
13403696
|
3M™
7000089596 |
4X-Large | 78 to 81 in. | 200 to 206 cm |
120.00 €
Pack of 20 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||
13443686
|
3M™
7000089590 |
Small | 64 to 67 in. | 164 to 170 cm |
120.00 €
Pack of 20 |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
13483686
|
3M™
7000089594 |
2X-Large | 73 to 76 in. | 186 to 194 cm |
120.00 €
Pack of 20 |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
13453686
|
3M™
7000089591 |
Medium | 66 to 69 in. | 167 to 176 cm |
120.00 €
Pack of 20 |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
13493686
|
3M™
7000089595 |
3X-Large | 76 to 78 in. | 194 to 200 cm |
120.00 €
Pack of 20 |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
13463686
|
3M™
7000089592 |
Large | 69 to 71 in. | 174 to 181 cm |
120.00 €
Pack of 20 |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
13473686
|
3M™
7000089593 |
X-Large | 70 to 74 in. | 179 to 187 cm |
120.00 €
Pack of 20 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
Description
CE Simple, Category 1 coverall is designed for domestic DIY use, light-duty industrial cleaning and maintenance applications, or environments where hygiene and cleanliness are important.- Excellent economic solution to keep the wearer clean and dry
- Suitable for non-hazardous applications
- Breathable and light-duty protection
- Strong, durable 45gsm polypropylene fabric
- Storm flap over zip
- Elasticated hood, cuffs, ankles, and waist
Specifications
General Purpose Coveralls | |
Polypropylene | |
Single-use | |
Elastic | |
2-panel | |
Serged | |
Category I | |
Light Industrial Cleaning, Maintenance |
Unisex | |
White | |
Elastic | |
Elastic | |
Front Zipper with Storm Flap | |
Non-sterile | |
4500W |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title