missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Height (English) | Quantity | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (English) | Quantity | Type | Price | Quantity & Availability | |||||
15446505
|
Thermo Scientific Chemicals
042628.KJ |
1 ft. | 5/Pk. | Vial | N/A | ||||||
15456505
|
Thermo Scientific Chemicals
042628.KM |
1 ft. | 25/Pk. | Vial |
49.04 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11405280
|
Thermo Scientific Chemicals
044530.KM |
2 ft. | 25/Pk. | Vial | N/A | ||||||
11415280
|
Thermo Scientific Chemicals
044530.KN |
2 in. | 100/Pk. | Vial |
143.72 €
Pack of 100 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
15402007
|
Thermo Scientific Chemicals
044531.KT |
Each | Vial Adapter | N/A | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Polystyrene vial with polyethylene slip-on cap; 1 in. long x 0.5 in. dia.
- Polystyrene vial with polyethylene slip-on cap; 2 in. long x 0.5 in. dia.
- Adapter for 2 in. long, 0.5 in. diameter vials
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Wig-L-Bug™ Accessories