Learn More
Thermo Scientific Chemicals Wig-L-Bug™ Accessories
For use with Digital Wig-L-Bug Mixing/Grinding Mill
Brand: Thermo Scientific Chemicals 044530.KM
45.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Polystyrene vial with polyethylene slip-on cap; 1 ft. long x 0.5 in. dia.
- Polystyrene vial with polyethylene slip-on cap; 2 ft. long x 0.5 in. dia.
- Adapter for 2 in. long, 0.5 in. diameter vials

Specifications
Polystyrene | |
25/Pk. | |
Digital Wig-L-Bug mixing/grinding mill |
2 ft. | |
Vial |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only