Learn More
Bel-Art™ Write-On™ Paper Label Tapes
Use pencil, ball-point pen, or solvent ink marker to create labels that remain legible under the most demanding laboratory conditions. Paper label tapes resist moisture and most solvents and are great for use in low temperature applications.
28.34 € - 219.00 €
Specifications
Autoclavable Autoclavable
Adhesive Type Rubber, Resin
Material Paper
Printable Sides Single-side Printable
Print Technology Ink Jet
View More Specs
Products 23
Product Code Brand Diameter (English) Length (English) Width (English) Color Price Quantity & Availability  
Product Code Brand Diameter (English) Length (English) Width (English) Color Price Quantity & Availability  
13455236
View Documents
Bel-Art™
F13488-0050
3 in. 40 yd. 0.5 in. Orange
70.82 €
Pack of 6
13465236
View Documents
Bel-Art™
F13487-0050
3 in. 40 yd. 0.5 in. Blue
121.00 €
Pack of 6
13405246
View Documents
Bel-Art™
F13487-0075
3 in. 40 yd. 0.75 in. Blue
188.00 €
Pack of 4
13435236
View Documents
Bel-Art™
F13484-0050
3 in. 40 yd. 0.5 in. Red
121.00 €
Pack of 6
13495236
View Documents
Bel-Art™
F13485-0075
3 in. 40 yd. 0.75 in. Yellow
199.00 €
Pack of 4
13455226
View Documents
Bel-Art™
F13480-0075
1 in. 15 yd. 0.75 in. White
28.34 €
Pack of 4
13475246
View Documents
Bel-Art™
F13487-0100
3 in. 40 yd. 1 in. Blue
98.40 €
Pack of 3
13415246
View Documents
Bel-Art™
F13486-0075
3 in. 40 yd. 0.75 in. Green
192.00 €
Pack of 4
13474166
View Documents
Bel-Art™
F13483-0050
3 in. 40 yd. 0.5 in. White
219.00 €
Pack of 6
10045180
View Documents
Bel-Art™
F13490-0075
3 in. 60 yd. 0.75 in. White
83.85 €
Pack of 4
13425246
View Documents
Bel-Art™
F13488-0075
3 in. 40 yd. 0.75 in. Orange
190.00 €
Pack of 4
13475236
View Documents
Bel-Art™
F13483-0075
3 in. 40 yd. 0.75 in. White
118.00 €
Pack of 4
13405236
View Documents
Bel-Art™
F13480-0100
1 in. 15 yd. 1 in. White
31.55 €
Pack of 3
13435246
View Documents
Bel-Art™
F13483-0100
3 in. 40 yd. 1 in. White
61.00 €
Pack of 3
13445246
View Documents
Bel-Art™
F13484-0100
3 in. 40 yd. 1 in. Red
58.83 €
Pack of 3
13465246
View Documents
Bel-Art™
F13486-0100
3 in. 40 yd. 1 in. Green
70.05 €
Pack of 3
13485236
View Documents
Bel-Art™
F13484-0075
3 in. 40 yd. 0.75 in. Red
188.00 €
Pack of 4
15196324
View Documents
Bel-Art™
F13463-0600
3 in. 15 yd. 0.75 in. Assorted
178.00 €
Pack of 6
13485246
View Documents
Bel-Art™
F13488-0100
3 in. 40 yd. 1 in. Orange
77.15 €
Pack of 3
13495246
View Documents
Bel-Art™
F13490-0050
3 in. 60 yd. 0.5 in. White
97.46 €
Pack of 6
13434720
View Documents
Bel-Art™
F13485-0050
3 in. 40 yd. 0.5 in. Yellow
84.30 €
Pack of 6
13455246
View Documents
Bel-Art™
F13485-0100
3 in. 40 yd. 1 in. Yellow
67.70 €
Pack of 3
13445236
View Documents
Bel-Art™
F13486-0050
3 in. 40 yd. 0.5 in. Green
72.80 €
Pack of 6
Description
Description
Use pencil, ball-point pen, or solvent ink marker to create labels that remain legible under the most demanding laboratory conditions. Paper label tapes resist moisture and most solvents and are great for use in low temperature applications.
  • Adheres to any clean surface, including PTFE fluoropolymer resin coated materials
  • Peels off without leaving a sticky residue
  • Pencil, ball-point pen, or solvent ink marker remain legible under the most demanding laboratory conditions
  • Colored paper with rubber resin adhesive resists moisture and most solvents
  • Withstands a maximum temperature of 157°C (315°F) for 30 minutes or 80°C (176°F) for 24 hours; can be used down to -80°C (-112°F)
  • Withstands a steam autoclaving cycle at 121°C (250°F) and gamma irradiation between 13.0 and 45.0kGy
Specifications
Specifications
Autoclavable
Paper
Ink Jet
Non-sterile
Rubber, Resin
Single-side Printable
Ultra Reflective Letter Label
For use in low temperature applications
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Bel-Art™ Write-On™ Paper Label Tapes