Learn More
ABEBA™ X-Light Black Lace Up Safety Shoes S2
ABEBA™ X-Light Black Lace Up Safety Shoes S2 with silver ions lining to reduce odor and a replaceable Active Comfort insole.
Brand:  ABEBA™ 7131027/46
 View more versions of this product
Product Code. 16328261
  • 113.00 € / 1 pair
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20345 : 2012, S2 SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant Mon PU outsole SRC
  • Good cushioning
  • Black washable microfiber
  • Silver ions lining
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Specifications
CE EN ISO 20345:2012 S2, SRC, ESD
Black
Microfiber
Slip and abrasion resistant, shock absorbing, ESD dissipative
Lace-up shoes
1 Pair
Water
Laces
Breathable lining with Silver ions
46
Polyurethane
Steel
CE EN ISO 20345:2012, DIN EN 61340
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
ABEBA™ X-Light Black Lace Up Safety Shoes S2 > Size: 46