missing translation for 'onlineSavingsMsg'
Learn More

Zinc neodecanoate, Zn 17.9-18.2%, Thermo Scientific Chemicals

91.15 € - 435.17 €

Chemical Identifiers

CAS 27253-29-8
Molecular Formula C20H40O4Zn
Molecular Weight (g/mol) 409.916
MDL Number MFCD00014384
InChI Key VADFQXONMAETJP-UHFFFAOYSA-N
Synonym zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
PubChem CID 117064711
IUPAC Name 7,7-dimethyloctanoic acid;zinc
SMILES CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11309687
View Documents
Thermo Scientific Alfa Aesar
041312.30
250 g
91.15 €
250g
Please to purchase this item. Need a web account? Register with us today!
11389677
View Documents
Thermo Scientific Alfa Aesar
041312.A1
1 kg
164.82 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11399677
View Documents
Thermo Scientific Alfa Aesar
041312.A7
5 kg
435.17 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zinc neodecanoate is used as a catalyst, lubricant additive and drier. Further it is used in the production of rubber and plastic products. In addition it serves as a polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

27253-29-8
409.916
VADFQXONMAETJP-UHFFFAOYSA-N
117064711
CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]
C20H40O4Zn
MFCD00014384
zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
7,7-dimethyloctanoic acid;zinc

Specifications

27253-29-8
102°C (216°F)
MFCD00014384
Immiscible with water.
CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]
409.916
407.89
Zinc neodecanoate
Brown
C20H40O4Zn
zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
VADFQXONMAETJP-UHFFFAOYSA-N
7,7-dimethyloctanoic acid;zinc
117064711
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 248-370-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only