Learn More
Zirconium dinitrate oxide hydrate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals
81.70 € - 587.00 €
Chemical Identifiers
CAS | 14985-18-3 |
---|---|
Molecular Formula | N2O7Zr |
Molecular Weight (g/mol) | 231.23 |
MDL Number | MFCD00149896 |
InChI Key | JWBWFTANBHYHBC-UHFFFAOYSA-N |
Synonym | zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis |
PubChem CID | 16211674 |
SMILES | [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O |
Description
Zirconium dintrate oxide hydrate is used to prepare CeO2-ZrO2 mixed oxide catalyst supports. It may be used as zirconium precursor in the preparation of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and Silica- and alumina-doped nanocrystalline ceria/zirconia (Ce0.5Zr0.5O2).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14985-18-3 | |
231.23 | |
JWBWFTANBHYHBC-UHFFFAOYSA-N | |
16211674 |
N2O7Zr | |
MFCD00149896 | |
zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis | |
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O |
Specifications
14985-18-3 | |
Powder | |
ZrO(NO3)2·xH2O | |
UN1477 | |
Soluble in alcohol | |
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O | |
231.23 | |
231.23 (Anhydrous) | |
Puratronic™ | |
(metals basis) |
White | |
N2O7Zr | |
MFCD00149896 | |
zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis | |
JWBWFTANBHYHBC-UHFFFAOYSA-N | |
(nitrooxy)(oxo)zirconio nitrate | |
16211674 | |
99.994% | |
Odorless | |
Zirconium dinitrate oxide hydrate |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 237-529-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only