missing translation for 'onlineSavingsMsg'
Learn More
1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%, Thermo Scientific Chemicals Product Code.: 15491596

1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%, Thermo Scientific Chemicals

Product Code. 15491596
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15491596

Brand: Thermo Scientific Alfa Aesar H54792.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-(2-Mesitylenesulfonyl)-3-Nitro-1H-1,2,4-Triazole is an efficient coupling reagent in the synthesis of oligonucleotides via phosphotriester approach.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Mesitylenesulfonyl)-3-Nitro-1H-1,2,4-Triazole is an efficient coupling reagent in the synthesis of oligonucleotides via phosphotriester approach.

Solubility
Soluble in Chloroform.

Notes
Store at 4°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74257-00-4
Molecular Formula C11H12N4O4S
Molecular Weight (g/mol) 296.30
MDL Number MFCD00009754
InChI Key SFYDWLYPIXHPML-UHFFFAOYSA-N
Synonym 1-mesitylsulfonyl-3-nitro-1h-1,2,4-triazole, 1-mesitylene-2-sulfonyl-3-nitro-1,2,4-triazole, msnt, 1-mesitylene-2-sulfonyl-3-nitro-1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonyl-1,2,4-triazole, 1-2-mesitylenesulfonyl-3-nitro-1h-1,2,4-triazole, 2,4,6-trimethylphenyl-3-nitro-1,2,4-triazol-1-yl sulfone, 3-nitro-1-2,4,6-trimethylbenzenesulfonyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonylShow MoreShow Less
PubChem CID 716901
IUPAC Name 3-nitro-1-(2,4,6-trimethylphenyl)sulfonyl-1,2,4-triazole
SMILES CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O

Specifications

Melting Point 134°C to 139°C
Quantity 1 g
Beilstein 4328921
Solubility Information Soluble in Chloroform.
Formula Weight 296.3
Percent Purity ≥99%
Chemical Name or Material 1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP30J004 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberH54792.03
Lot NumberP21I032 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH54792.06,H54792.03
Lot NumberQ25C020 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54792.06,H54792.03
Lot NumberBC240501 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54792.06,H54792.03
Lot NumberN21E055 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54792.06,H54792.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.