missing translation for 'onlineSavingsMsg'
Learn More
1,7-Octadiyne, 98%, Thermo Scientific Chemicals Product Code.: 11452508

1,7-Octadiyne, 98%, Thermo Scientific Chemicals

Product Code. 11452508
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11452508

Brand: Thermo Scientific Alfa Aesar A15829.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,7-Octadiyne is an useful bifunctional chain extension unit. It participates in the formation of uranium(IV) vinyl complexes. It reacts with frustrated Lewis pair P(o-tolyl)3/B(C6F5)3 by acetylene C-C coupling to yield the zwitterionic product. It undergoes the reductive cyclization during reaction with Re2Cl4(?-dppm)2 (dppm = Ph2PCH2PPh2) and affords quadruply bonded dirhenium(III) complex Re2Cl3(?,η2-C8H7)(?-dppm)2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Octadiyne is an useful bifunctional chain extension unit. It participates in the formation of uranium(IV) vinyl complexes. It reacts with frustrated Lewis pair P(o-tolyl)3/B(C6F5)3 by acetylene C-C coupling to yield the zwitterionic product. It undergoes the reductive cyclization during reaction with Re2Cl4(μ-dppm)2 (dppm = Ph2PCH2PPh2) and affords quadruply bonded dirhenium(III) complex Re2Cl3(μ,η2-C8H7)(μ-dppm)2.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871-84-1
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008580
InChI Key DSOJWVLXZNRKCS-UHFFFAOYSA-N
Synonym 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ciShow MoreShow Less
PubChem CID 70099
IUPAC Name octa-1,7-diyne
SMILES C#CCCCCC#C

Specifications

Density 0.81
Boiling Point 135°C to 136°C
Flash Point 23°C (73°F)
Odor Recognizable
Refractive Index 1.446
Quantity 10 g
UN Number UN3295
Beilstein 1697275
Solubility Information Not miscible or difficult to mix.
Formula Weight 106.17
Percent Purity 98%
Chemical Name or Material 1,7-Octadiyne
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242991 Certificate TypeCertificate of Analysis Date31/08/2022 Catalog NumberA15829.09,A15829.18
Lot Number10240001 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15829.09,A15829.18
Lot Number10131969 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15829.09,A15829.18
Lot NumberUSLF004918 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15829.09,A15829.18
Lot Number10157481 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15829.09,A15829.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,7-Octadiyne, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.