missing translation for 'onlineSavingsMsg'
Learn More
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech., Thermo Scientific Chemicals Product Code.: 11324344

2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech., Thermo Scientific Chemicals

Product Code. 11324344
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11324344

Brand: Thermo Scientific Alfa Aesar L00875.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-02-0
Molecular Formula C4HF7O
Molecular Weight (g/mol) 198.04
MDL Number MFCD00039731
InChI Key IQJZGNJYXIIMGP-UHFFFAOYSA-N
Synonym heptafluorobutyraldehyde, butanal, heptafluoro, butyraldehyde, heptafluoro, heptafluorobutyraldehyde, tech., perfluorobutanal, perfluorobutyraldehyde, c3f7cho, heptafluoro-n-butyraldehyde, butanal,2,2,3,3,4,4,4-heptafluoroShow MoreShow Less
PubChem CID 67809
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanal
SMILES FC(F)(F)C(F)(F)C(F)(F)C=O

Specifications

Refractive Index 1.32
Quantity 1 g
Beilstein 1367365
Formula Weight 198.04 (Anhydrous)
Grade Technical
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250976 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL00875.03,L00875.06
Lot Number10246611 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog NumberL00875.03,L00875.06
Lot Number10246657 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog NumberL00875.03,L00875.06
Lot NumberB06S048 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00875.03,L00875.06
Lot NumberUSLF004039 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00875.03,L00875.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.