missing translation for 'onlineSavingsMsg'
Learn More
2,3-Benzanthracene, 98%, Thermo Scientific Chemicals Product Code.: 11954451

2,3-Benzanthracene, 98%, Thermo Scientific Chemicals

Product Code. 11954451
5 g, Glass bottle
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11954451

Brand: Thermo Scientific Acros 105260050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-24-0
Molecular Formula C18H12
Molecular Weight (g/mol) 228.29
InChI Key IFLREYGFSNHWGE-UHFFFAOYSA-N
Synonym naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183Show MoreShow Less
PubChem CID 7080
ChEBI CHEBI:32600
IUPAC Name tetracene
SMILES C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1

Specifications

Melting Point >300°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 05,IV,2545
Merck Index 15,6454
Solubility Information Solubility in water: insoluble. Other solubilities: difficulty soluble in most solvents,soluble in hot xylene
Formula Weight 228.29
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Benzanthracene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0404986 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog Number105260010,105265000,105260050
Lot Number1237884 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number105260010,105265000,105260050
Lot Number1232310 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number105260010,105265000,105260050
Lot Number1227491 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number105260010,105265000,105260050
Lot Number1222442 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number105260010,105265000,105260050
compliance-icons
Product Identifier
  • 2,3-Benzanthracene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Benzanthracene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.