missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dibromoaniline, 97%, Thermo Scientific Chemicals Product Code.: 11409416

2,5-Dibromoaniline, 97%, Thermo Scientific Chemicals

Product Code. 11409416
25 g
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11409416

Brand: Thermo Scientific Alfa Aesar A19114.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3638-73-1
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00007636
InChI Key WRTAZRGRFBCKBU-UHFFFAOYSA-N
Synonym 2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoanilineShow MoreShow Less
PubChem CID 77198
IUPAC Name 2,5-dibromoaniline
SMILES C1=CC(=C(C=C1Br)N)Br

Specifications

Melting Point 51°C to 56°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
UN Number UN2811
Beilstein 1817548
Formula Weight 250.93
Percent Purity 98%
Chemical Name or Material 2,5-Dibromoaniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG9985B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19114.14,A19114.22,A19114.06
Lot Number10212313 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19114.14,A19114.22,A19114.06
Lot Number10179921 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19114.14,A19114.22,A19114.06
Lot NumberH2363A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19114.14,A19114.22,A19114.06
Lot Number10229257 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19114.14,A19114.22,A19114.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dibromoaniline, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.