missing translation for 'onlineSavingsMsg'
Learn More
2-Hexanone, 98%, Thermo Scientific Chemicals Product Code.: 11349609

2-Hexanone, 98%, Thermo Scientific Chemicals

Product Code. 11349609
10 g
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11349609

Brand: Thermo Scientific Alfa Aesar A14867.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (±)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (+/-)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.

Solubility
Miscible with acetone, ethanol and ether. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-78-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009482
InChI Key QQZOPKMRPOGIEB-UHFFFAOYSA-N
Synonym 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methylShow MoreShow Less
PubChem CID 11583
IUPAC Name hexan-2-one
SMILES CCCCC(C)=O

Specifications

Melting Point -56°C
Density 0.811
Boiling Point 126°C to 127°C
Flash Point 23°C (73°F)
Odor Pungent
Refractive Index 1.401
Quantity 10 g
UN Number UN1224
Beilstein 1737676
Merck Index 14,6033
Solubility Information Miscible with acetone,ethanol and ether. Slightly miscible with water.
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material 2-Hexanone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462053 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14867.36,A14867.22,A14867.09,A1486701,A14867.18
Lot NumberA0442805 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog NumberA14867.36,A14867.22,A14867.09,A14867.18
Lot Number10181162 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14867.36,A14867.22,A14867.09,A14867.18
Lot Number10146444 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14867.36,A14867.22,A14867.09,A14867.18
Lot Number10169263 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14867.36,A14867.22,A14867.09,A14867.18
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Hexanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2-Hexanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Hexanone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.