Learn More
Endecotts™ Stainless Steel Test Sieve, 200 mm dia.
Full height test sieves provide all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Test Sieves, 200 mm dia. comply with ISO 3310-1 / BS410 standards.
136.00 € - 800.00 €
Specifications
Certifications/Compliance | ISO 3310-1 / BS410 |
---|---|
Diameter (Metric) Outer | 200 mm |
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
Product Code | Brand | Pore Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | ||||||
11597474
|
Endecotts™
200SIW1.18 |
1.18 mm |
136.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
11567474
|
Endecotts™
200SIW1.70 |
1.7 mm |
136.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
10644415
|
Endecotts™
200SIW1.40 |
1.4 mm |
136.00 €
Each |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
12991985
|
Endecotts™
200SIW.150 |
150 μm | N/A | N/A | N/A | |||||
10092184
|
Endecotts™
200SIW.180 |
180 μm |
147.00 €
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
11547434
|
Endecotts™
200SIW106. |
106 mm |
173.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11634762
|
Endecotts™
200SIW.125 |
125 μm |
136.00 €
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
11527474
|
Endecotts™
200SIW2.36 |
2.36 mm |
136.00 €
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
11654762
|
Endecotts™
200SIW1.00 |
1 mm |
136.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
10022044
|
Endecotts™
200SIW.020 |
20 μm |
800.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
12065240
|
Endecotts™
200SIW.212 |
212 μm |
136.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
10566544
|
Endecotts™
200SIW.250 |
250 μm |
147.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
10265254
|
Endecotts™
200SIW2.00 |
2 mm |
143.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
11587464
|
Endecotts™
200SIW3.35 |
3.35 mm |
136.00 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
10133734
|
Endecotts™
200SIW.032 |
32 μm |
353.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
11557444
|
Endecotts™
200SIW35.5 |
35.5 mm |
173.00 €
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
11577444
|
Endecotts™
200SIW28.0 |
28 mm |
173.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
10769045
|
Endecotts™
200SIW.038 |
38 μm |
208.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
10114214
|
Endecotts™
200SIW.045 |
45 μm |
191.00 €
Each |
Estimated Shipment: 09-06-2025
Log in to see stock. |
||||||
10585574
|
Endecotts™
200SIW40.0 |
4 mm |
173.00 €
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
11527444
|
Endecotts™
200SIW50.0 |
50 mm |
173.00 €
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||||
10613865
|
Endecotts™
200SIW.053 |
53 μm |
164.00 €
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
12971985
|
Endecotts™
200SIW.600 |
600 μm |
136.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
11587434
|
Endecotts™
200SIW71.0 |
71 mm |
173.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
10443394
|
Endecotts™
200SIW.850 |
850 μm |
136.00 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10134214
|
Endecotts™
200SIW.075 |
75 μm |
159.00 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
10022524
|
Endecotts™
200SIW.090 |
90 μm |
136.00 €
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
10452084
|
Endecotts™
200SIW.063 |
63 μm |
146.00 €
Each |
Estimated Shipment: 01-05-2025
Log in to see stock. |
||||||
10235494
|
Endecotts™
200SIW.300 |
300 μm |
136.00 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
12981985
|
Endecotts™
200SIW.425 |
425 μm |
147.00 €
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
Description
Full height test sieves provide all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Test Sieves, 200 mm dia. comply with ISO 3310-1 / BS410 standards.
- Stainless steel precision frame (ensures consistent nestability)
- Stainless steel mesh
- Precise aperture in accordance with ISO 3310-1 / BS410
- Natural fillet (free flowing of sample)
- Totally sealed (no crevice to lose material)
- Evenly tensioned mesh (ensures accurate analysis)
- Safe edge (big radius makes it comfortable to handle)
- Serial number (ensures full traceability)
Specifications
ISO 3310-1 / BS410 | |
200 mm | |
Stainless Steel | |
Stainless Steel |