missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2H-Perfluoro-5-methyl-3,6-dioxanonane, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
CAS | 3330-14-1 |
Melting Point | -122°C |
Density | 1.659 |
Boiling Point | 103°C to 105°C |
Flash Point | None |
Molecular Formula | C8HF17O2 |
Refractive Index | 1.3005 |
MDL Number | MFCD00054714 |
Quantity | 5 g |
Synonym | 2h-perfluoro-5-methyl-3,6-dioxanonane, 1,1,1,2,2,3,3-heptafluoro-3-1,1,1,2,3,3-hexafluoro-3-1,2,2,2-tetrafluoroethoxy propan-2-yl oxy propane, propane, 1-1-difluoro 1,2,2,2-tetrafluoroethoxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2,3,3,3-heptafluoro, cf3cfhocf2cf cf3 ocf2cf2cf3, 2h-perfluoro 5-methyl-3,6-dioxanonane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy-3-1,2,2,2-tetrafluoroethoxy propane, 1,1,1,2,4,4,5,7,7,8,8,9,9,9-tetradecafluoro-5-trifluoromethyl-3,6-dioxanonane |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2H-Perfluoro-5-methyl-3,6-dioxanonane, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction