missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-fluorobenzyl bromide, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Bromo-5-fluorobenzyl bromide is employed as a intermediate for pharmaceutical and chemical research.
Solubility
Slightly soluble in water (2.9 g/L) (25°C).
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
3-Bromo-5-fluorobenzyl bromide is employed as a intermediate for pharmaceutical and chemical research.
Solubility
Slightly soluble in water (2.9 g/L) (25°C).
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Specifications
Specifications
Quantity | 1 g |
UN Number | UN3261 |
Solubility Information | Slightly soluble in water (2.9g/L) (25°C). |
Formula Weight | 267.92 |
Percent Purity | 95% |
Chemical Name or Material | 3-Bromo-5-fluorobenzyl bromide |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Bromo-5-fluorobenzyl bromide, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction