missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™
43.50 € - 328.00 €
Chemical Identifiers
CAS | 4926-47-0 |
---|---|
Molecular Formula | C7H5BrN2 |
Molecular Weight (g/mol) | 197.035 |
InChI Key | APYSHMNJHJRIDR-UHFFFAOYSA-N |
PubChem CID | 327951 |
IUPAC Name | 3-bromoimidazo[1,2-a]pyridine |
SMILES | C1=CC2=NC=C(N2C=C1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10334622
|
Thermo Scientific Maybridge
CC25410DA |
1 g |
43.50 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10326574
|
Thermo Scientific Maybridge
CC25410DE |
5 g |
179.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10342005
|
Thermo Scientific Maybridge
CC25410EA |
10 g |
328.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
4926-47-0 | |
197.035 | |
327951 | |
C1=CC2=NC=C(N2C=C1)Br |
C7H5BrN2 | |
APYSHMNJHJRIDR-UHFFFAOYSA-N | |
3-bromoimidazo[1,2-a]pyridine |
Specifications
4926-47-0 | |
Amber glass bottle | |
3-bromoimidazo 1,2-a pyridine, 3-bromo-imidazo 1,2-a pyridine, 3-bromopyrimidazole, imidazo 1,2-a pyridine, 3-bromo, 3-bromoimidazolo 1,2-a pyridine, 3-bromoh-imidazo 1,2-a pyridine, pubchem17183, acmc-209kfj, buttpark 96\50-11, imidazo 1,2-a pyridine,3-bromo | |
C1=CC2=NC=C(N2C=C1)Br | |
197.035 | |
197.03 | |
3-Bromoimidazo[1, 2-a]pyridine |
97% | |
C7H5BrN2 | |
APYSHMNJHJRIDR-UHFFFAOYSA-N | |
3-bromoimidazo[1,2-a]pyridine | |
327951 | |
97% |