missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 3-Iodo-L-tyrosine, 97% Product Code.: 10285160

Thermo Scientific Chemicals 3-Iodo-L-tyrosine, 97%

Product Code. 10285160
1 g, Glass Bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10285160

Brand: Thermo Scientific Chemicals 122440010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-78-0
Molecular Formula C9H10INO3
Molecular Weight (g/mol) 307.09
MDL Number MFCD00002608
InChI Key UQTZMGFTRHFAAM-JLDDOWRYNA-N
Synonym 3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanineShow MoreShow Less
PubChem CID 439744
ChEBI CHEBI:27847
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O

Specifications

Melting Point 196.0°C to 198.0°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Specific Rotation − 3.18
Merck Index 15, 5090
Solubility Information Solubility in water: .. Other solubilities: soluble in 15 parts boiling water
Water 1% max. (K.F.)
Quantity 1 g
Formula Weight 307.08
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Iodo-L-tyrosine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0377636 Certificate TypeCertificate of Analysis Date12/10/2023 Catalog Number122440250,122440050,122440010
Lot NumberA0439770 Certificate TypeCertificate of Analysis Date22/03/2022 Catalog Number122440250,122440050,122440010
Product Identifier
  • 3-Iodo-L-tyrosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 3-Iodo-L-tyrosine, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.