Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve with Square Holes
Brand: RETSCH 60.138.080000
Specifications
1Each | |
Test Sieve | |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Stainless Steel |
80 mm | |
ISO 3310/2 | |
40mm | |
305 mm | |
Stainless Steel |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.