missing translation for 'onlineSavingsMsg'
Learn More
3M™ Gray Protective Coverall 4570 Product Code.: 15799027
Estimated Shipment: 12-05-2025
to see stock.
Add to basket

3M™ Gray Protective Coverall 4570

Product Code. 15799027
X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy

Product Code. 15799027

Brand: 3M™ 7000089690

Estimated Shipment: 12-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Protective Coverall 4570 helps protect against a range of chemical jets and sprays, and has been certified to Type 3/4/5/6 levels of protection.

3M™ Protective Coverall 4570 offers chemical hold out and mechanical strength for workers in environments with chemical exposure. The suit helps protect against hazardous dusts (Type 5), light liquid splashes (Type 6), low pressure liquid sprays (Type 4) and high pressure liquid jets (Type 3). It is also certified to offer protection against radioactive particles (EN 1073-2) and (EN 14126). This suit achieves the highest class performance on all tests in EN 14126 and passes ASTM F1670 and F1671 for helping to protect against biological contaminants.

  • A Versatile Protective Suit for many Applications
  • Protective coverall helps protect against a range of chemical jets and sprays
  • Heavy-duty fabric with a soft anti-static treated inner layer offers protection and convenience
  • Stitched seams are sealed with a 5-layer chemically protective tape
  • Two storm flaps create a double seal for extra protection
  • Large thumb loops and zipper pulls offer convenience while wearing gloves
  • Knit polyester cuffs for user comfort
  • Anti-static, low-linting, microporous
  • Elastic around the head for secure fit
  • Elastic around the face to ensure secure fit
  • Elastic at the calf to provide closure around the top of the footwear for protection against contaminants
  • Elastic around the ankle for protection against contaminants and enhanced fit
  • Universal size with ties to provide a good fit for a wide range of users
  • Lightweight and comfortable to wear
TRUSTED_SUSTAINABILITY

Specifications

Size X-Large
Material Polyethylene
Color Gray
Waist Style Elastic
Cuff Style Elastic
Hood Type 3-panel
Closure Type Front Zipper with Double Storm Flap
Seam Type Taped
Protection Class 3
Latex Free Yes
Model 4570-XL
No. of Pockets None
Size (English) Chest 43 – 45in.
Size (Metric) Chest 108 – 115 cm
For Use With (Application) Chemical mixing and handling, chemical clean-up, hazardous waste remediation, decontamination, tank cleaning, handling toxic powders, infective agents
Height (English) 70 to 74 in.
Show More Show Less
  • 3M Safety Solutions - Protective coveralls overview
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      3M Safety Solutions - Protective coveralls overview

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    3M™ Gray Protective Coverall 4570 >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.