missing translation for 'onlineSavingsMsg'
Learn More
4-Amino-2-methoxybenzoic acid, 97%, Thermo Scientific Chemicals Product Code.: 11397239

4-Amino-2-methoxybenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11397239
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11397239

Brand: Thermo Scientific Alfa Aesar H27419.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2486-80-8
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00114479
InChI Key OLJXRTRRJSMURJ-UHFFFAOYSA-N
Synonym 4-amino-o-anisic acid, 4-amino-2-methoxy-benzoic acid, 4-amino-2-methoxybenzenecarboxylic acid, 2-methoxy-4-amino-benzoic acid, 4-amino-2-methoxybenzoicacid, 4-azanyl-2-methoxy-benzoic acid, benzoic acid, 4-amino-2-methoxy, 2-methoxy-4-aminobenzoic acid, acmc-20aicb, pubchem4934
PubChem CID 75599
IUPAC Name 4-amino-2-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)N)C(=O)O

Specifications

Melting Point ∽150°C (decomposition)
Quantity 25 g
Formula Weight 167.16
Percent Purity 97%
Chemical Name or Material 4-Amino-2-methoxybenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10142173 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27419.14,H27419.06
Lot Number10135406 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27419.14,H27419.06
Lot Number10173039 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27419.14,H27419.06
Lot Number10206301 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27419.14,H27419.06
Lot Number10152760 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27419.14,H27419.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Amino-2-methoxybenzoic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.