missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 4-Amino-DL-phenylalanine Product Code.: 15476839

Thermo Scientific Chemicals 4-Amino-DL-phenylalanine

Product Code. 15476839
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15476839

Brand: Thermo Scientific Chemicals J65331.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2922-41-0
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
MDL Number MFCD00007917
InChI Key CMUHFUGDYMFHEI-UHFFFAOYSA-N
Synonym 4-amino-dl-phenylalanine, 2-amino-3-4-aminophenyl propanoic acid, p-amino-dl-phenylalanine, p-aminophenylalanine, phenylalanine, 4-amino, 4-aminophenylalanine, h-dl-phe 4-nh2-oh, p-aminophenyl alanine, p-amino-dl-phenylalanine hydrate, l-2-amino-3-4-aminophenyl propanoic acidShow MoreShow Less
PubChem CID 95174
IUPAC Name 2-amino-3-(4-aminophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)N

Specifications

Melting Point 228°C to 230°C
Color White to Brown
Solubility Information Soluble in water
Quantity 1 g
Formula Weight 180.21
Physical Form Powder
Chemical Name or Material 4-Amino-DL-phenylalanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06K043 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberJ65331.06,J65331.03
Lot NumberN19K031 Certificate TypeCertificate of Analysis Date06/03/2024 Catalog NumberJ65331.06,J65331.03
Lot NumberZ13D001 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ65331.06,J65331.03
Lot NumberI04Y053 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ65331.06,J65331.03
Lot NumberU17C027 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ65331.06,J65331.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 4-Amino-DL-phenylalanine >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.