missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenol, 98%, Thermo Scientific Chemicals

Product Code. 11430104
Click to view available options
Quantity:
50 g
250 g
1000 g
5000 g

4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.

Solubility
Soluble in acetonitrile, ethyl acetate, acetone, hot water, dimethylsulfoxide and alkalies. Slightly soluble in toluene, diethyl ether, ethanol, trifluoroacetic acid and cold water. Insoluble in benzene and chloroform.

Notes
Incompatible with acids, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-30-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00007869
InChI Key PLIKAWJENQZMHA-UHFFFAOYSA-N
Synonym p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
PubChem CID 403
ChEBI CHEBI:17602
IUPAC Name 4-aminophenol
SMILES C1=CC(=CC=C1N)O

Specifications

Melting Point 186°C to 190°C
Density 1.29
Boiling Point 284°C (decomposition)
Flash Point 195°C (383°F)
Assay Percent Range 98%
Quantity 250 g
UN Number UN2512
Beilstein 385836
Sensitivity Air and light sensitive
Merck Index 14,462
Solubility Information Soluble in acetonitrile,ethyl acetate,acetone,hot water,dimethylsulfoxide and alkalies. Slightly soluble in toluene,diethyl ether,ethanol,trifluoroacetic acid and cold water. Insoluble in benzene and chloroform.
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 4-Aminophenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Aminophenol
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H341-Suspected of causing genetic defects.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.