missing translation for 'onlineSavingsMsg'
Learn More
4-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals Product Code.: 10377170

4-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals

Product Code. 10377170
100 mL, Glass bottle
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10377170

Brand: Thermo Scientific Acros 150191000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-01-0
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
InChI Key RKIDDEGICSMIJA-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcbocShow MoreShow Less
PubChem CID 8501
ChEBI CHEBI:60716
IUPAC Name 4-chlorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)Cl

Specifications

Melting Point 12.0°C to 14.0°C
Color Colorless
Density 1.3700g/mL
Boiling Point 222.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4COCl
Refractive Index 1.5770 to 1.5790
Quantity 100 mL
Beilstein 09,341
Specific Gravity 1.37
Solubility Information Solubility in water: hydrolyses. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 175.01
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzoyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0437774 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number150195000,150191000,150190025
Lot NumberA0462738 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number150195000,150191000,150190025
Lot NumberA0450028 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog Number150195000,150191000,150190025
Lot NumberA0437774 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number150195000,150190050,150191000,150190025
Lot NumberA0406740 Certificate TypeCertificate of Analysis Date09/04/2021 Catalog Number150195000,150190050,150191000,150190025
compliance-icons
Product Identifier
  • 4-Chlorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.