missing translation for 'onlineSavingsMsg'
Learn More
(4-Hydroxymethyl-benzyl)-carbamic acid tert-butyl ester, 97%, Thermo Scientific™ Product Code.: 10035692

(4-Hydroxymethyl-benzyl)-carbamic acid tert-butyl ester, 97%, Thermo Scientific™

Product Code. 10035692
Amber Glass Bottle; 1g
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy

Product Code. 10035692

Brand: Thermo Scientific Maybridge MO07285DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 123986-64-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C13H19NO3
MDL Number MFCD06411553
Quantity 1g
Synonym tert-butyl 4-hydroxymethyl benzylcarbamate, 4-hydroxymethyl-benzyl-carbamic acid tert-butyl ester, tert-butyl n-4-hydroxymethyl phenyl methyl carbamate, n-boc-4-aminomethyl benzyl alcohol, n-boc-4-hydroxymethyl benzylamine, tert-butyl 4-hydroxymethyl benzyl carbamate, 4-aminomethyl benzyl alcohol, n-boc protected, 4-tert-butoxycarbonylaminomethyl benzyl alcohol, 4-n-tert-butoxycarbonyl aminomethyl-1-phenylmethanol
InChI Key KUEPOWVQABAWRK-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)CO
IUPAC Name tert-butyl N-[[4-(hydroxymethyl)phenyl]methyl]carbamate
Molecular Weight (g/mol) 237.299
PubChem CID 2794813
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material (4-Hydroxymethyl-benzyl)-carbamic acid tert-butyl ester
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(4-Hydroxymethyl-benzyl)-carbamic acid tert-butyl ester, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.