missing translation for 'onlineSavingsMsg'
Learn More
5,7-Dimethylisatin, 96%, Thermo Scientific Chemicals Product Code.: 11317716

5,7-Dimethylisatin, 96%, Thermo Scientific Chemicals

Product Code. 11317716
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11317716

Brand: Thermo Scientific Alfa Aesar L11332.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5,7-Dimethylisatin is used to produce cyano-(5,7-dimethyl-2-oxo-1,2-dihydro-indol-3-ylidene)-acetic acid ethyl ester by knoevenagel condensation with cyanoacetic acid ethyl ester. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,7-Dimethylisatin is used to produce cyano-(5,7-dimethyl-2-oxo-1,2-dihydro-indol-3-ylidene)-acetic acid ethyl ester by knoevenagel condensation with cyanoacetic acid ethyl ester. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39603-24-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00047219
InChI Key HFZSCCJTJGWTDZ-UHFFFAOYSA-N
Synonym 5,7-dimethylisatin, 5,7-dimethylindoline-2,3-dione, 5,7-dimethyl-indole-2,3-dione, 5,7-dimethyl-2,3-dihydro-1h-indole-2,3-dione, 1h-indole-2,3-dione, 5,7-dimethyl, indole-2,3-dione, 5,7-dimethyl, 1h-indole-2,3-dione, 5,7-dimethyl-9ci, acmc-20a4zq, isatin-based compound, 48, 1h-indole-2,3-dione,5,7-dimethylShow MoreShow Less
PubChem CID 38296
IUPAC Name 5,7-dimethyl-1H-indole-2,3-dione
SMILES CC1=CC(=C2C(=C1)C(=O)C(=O)N2)C

Specifications

Melting Point 244°C to 248°C
Quantity 5 g
Beilstein 143679
Solubility Information Insoluble in water.
Formula Weight 175.19
Percent Purity 96%
Chemical Name or Material 5,7-Dimethylisatin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251675 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog NumberL11332.14,L11332.06
Lot Number10197013 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog NumberL11332.14,L11332.06
Lot NumberD06S026 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL11332.14,L11332.06
Lot Number10192770 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11332.14,L11332.06
Lot NumberJ08S028 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11332.14,L11332.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5,7-Dimethylisatin, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.