Learn More
Thermo Scientific™ Acclaim™ AmG C18 HPLC Columns
Meet today’s demands for aminoglycoside antibiotic analyses, including drug purity characterization & quantification. They have unique tolerance to harsh conditions.
Brand: Thermo Scientific™ 088755
986.43 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Get a high-performance, reversed-phase columns for aminoglycoside analysis. The robust and rugged Thermo Scientific™ Acclaim™ AmG C18 HPLC column provides exceptional tolerance required for aminoglycosides ion-pairing chromatographic separations. These silica-based C18 columns are optimized for various aminoglycoside antibiotic analyses, including drug purity characterization and quantification, therapeutic drug monitoring, and residual control testing in different matrices. These columns are stable at low pH, high temperature, organic solvents, and aqueous mobile phases.
Highly Stable for Aminoglycoside AnalysesThe Acclaim AmG C18 column is specifically designed for ion-pairing reversed-phase HPLC (IP-RPLC) analysis of aminoglycoside antibiotics using volatile perfluorinated carboxylic acids as the ion-pairing reagent, such as 100 mM trifluoroacetic acid (TFA). The Acclaim AmG C18 stationary phase is based on a proprietary C18 bonding technology that provides extremely high stability towards acidic conditions. This specialty column exhibits excellent selectivity and high resolution for the aminoglycoside antibiotics analysis. In addition, it is easy to use because simple aqueous mobile phase (e.g. 100 mM TFA) without organic solvent and pH adjusting buffer is sufficient in most cases.
- Excellent selectivity for aminoglycosides
- Superior tolerance towards acidic conditions
- High efficiency and throughput
- Ease of use
The column can be used with UV, CAD, ELSD detectors

Specifications
Analytical Column | |
0.425 mL/min | |
8000 psi (550 bar) | |
0.5 to 10 | |
Each | |
L1 | |
Acclaim | |
Aminoglycoside Analysis | |
Responsibly packaged |
3 mm | |
150 mm | |
3 μm | |
120 Å | |
<80°C | |
C18 | |
Spherical, High Purity Silica | |
Reversed Phase |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.