missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AIM V™ Medium, liquid (research grade)
GREENER_CHOICE

AIM V™ Medium, liquid (research grade)

Brand:  Gibco™ 12055083

 View more versions of this product

Product Code. 10297582

  • 174.00 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Contains L-glutamine, streptomycin sulfate at 50μg/mL, and gentamicin sulfate at 10μg/mL.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes
Serum Free Medium (SFM)
AIM V™
Room Temperature
Research Grade
Mammalian Cell Culture
Liquid
Glutamine, Phenol Red, HSA
1000 mL
Streptomycin Sulfate, Gentamicin
Store in refrigerator (2–8°C) and protect from light.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2953883 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number12055083,12055091
Lot Number2366186 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12055083,12055091
Lot Number2390837 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12055083,12055091
Lot Number2451531 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12055083,12055091
Lot Number2451541 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12055083,12055091
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ AIM V™ Medium, liquid (research grade) > Quantity: 1000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.