Learn More
Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical
For two-component system. Water equivalent: 5.0mg H2O/mL
Brand: Fisher Chemical K/2200/15
168.64 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description

Specifications
Aqualine™ Titrant 5 | |
Karl Fischer Reagent | |
1 L | |
64°C | |
Liquid | |
CH4O | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol |
Colorless | |
Pyridine free | |
5.0mg of water/mL min. | |
Amber glass bottle | |
> 1.0 | |
MFCD00004595 | |
CO | |
32.04 |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Aqualin Titrant 5
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Methyl alcohol, Iodine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.