missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 647 Tyramide SuperBoost™ Kit, goat anti-mouse IgG

Features poly-HRP-conjugated goat anti-mouse IgG secondary antibody. Alexa Fluor™ 647 Tyramide SuperBoost™ Kit combine brightness of AlexaFluor dyes with high signal amplification of a poly-HRP-mediated tyramide labeling reaction to produce se

Brand:  Invitrogen™ B40916

492.48 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 15699240

  • 648.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).

Features:

  • High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
  • Simple protocol and detection using standard filters
  • Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
  • Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments

Benefits:

  • Enhancement of signal using Alexa Fluor tyramides: Utilize Alexa Fluor tyramides, which react with HRP to ultimately deposit bright and photostable Alexa Fluor dye on surrounding proteins and other similar molecules
  • Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
  • Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
  • Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals

Shelf Life:

  • Stable for 6 months after receiving
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alexa Fluor 647
1 kit sufficient for 150 microscope slides (18 x 18 mm), containing: Blocking buffer (1X), 22.5 mL
  • Poly-HRP-conjugated goat anti-mouse secondary antibody (1X), 22.5 mL
  • Alexa Fluor tyramide reagent
  • Hydrogen perox
  • SuperBoost™
    150 slides
    Approved for shipment at Room Temperature or on Wet Ice
    Tyramide Kit
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number2984243 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog NumberB40916
    Lot Number2907361 Certificate TypeCertificate of Analysis Date24/05/2024 Catalog NumberB40916
    Lot Number2770800 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog NumberB40916
    Lot Number2738297 Certificate TypeCertificate of Analysis Date25/07/2023 Catalog NumberB40916
    Lot Number2585988 Certificate TypeCertificate of Analysis Date20/02/2023 Catalog NumberB40916
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Invitrogen™ Alexa Fluor™ 647 Tyramide SuperBoost™ Kit, goat anti-mouse IgG > Alexa Fluor 647 Tyramide SuperBoost Kit, Goat anti-mouse IgG

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.