missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ Antibody Labeling Kits

Multiplex with brighter, photostable, Alexa Fluor Antibody Labeling Kits without the intramolecular quenching. Conjugate antibodies to 9 available fluorophores for flow cytometry, ICC, IHC, IF, ELISA or HCS. Each kit contains enough dye to conjugate 100 μg per reaction with 5 reactions per kit.

Brand:  Molecular Probes™ A20188

350.36 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


437.95 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 13305654

  • 478.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Labeling Kit
Alexa Fluor™ 680 Antibody Labeling Kit comes with 5 vials of reactive dye, sodium bicarbonate, purification resin, spin columns, and collections tubes.

Store at 2°C to 8°C, protected from light. Component A may be stored from -20°C to +8°C.
680/700 nm
Conjugation-based
Room Temperature
Amine
Alexa Fluor 680
Antibodies
5 Reactions kit
Fluorescence
Fluorescent Imager
100 μg
Alexa Fluor™
DMSO (Dimethylsulfoxide)
Near-IR
Alexa Fluor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3102003 Certificate TypeCertificate of Analysis Date11/01/2025 Catalog NumberA20188
Lot Number2743086 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog NumberA20188
Lot Number2532516 Certificate TypeCertificate of Analysis Date01/03/2023 Catalog NumberA20188
Lot Number2488972 Certificate TypeCertificate of Analysis Date21/07/2022 Catalog NumberA20188
Lot Number2441322 Certificate TypeCertificate of Analysis Date17/03/2022 Catalog NumberA20188
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Alexa Fluor™ Antibody Labeling Kits > Alexa Fluor™ 680

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.